missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ S100 Monoclonal Antibody (8B10)
GREENER_CHOICE

Product Code. 11571400
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11571400 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571400 Supplier Invitrogen™ Supplier No. MA122734

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-22734 detects S100 in human samples.

S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen S100
Applications ELISA, Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8B10
Concentration 4.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene S100B
Gene Accession No. P04271, P23297
Gene Alias AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P
Gene Symbols S100A1, S100B
Host Species Mouse
Immunogen Full length protein (Human).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6271, 6285
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.