missing translation for 'onlineSavingsMsg'
Learn More

S100 calcium binding protein A14 Antibody (S100A14/7401), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen S100 calcium binding protein A14
Clone S100A14/7401
Concentration 0.2 mg/ml
Dilution ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL
Applications ELISA, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18684744
View Documents
Novus Biologicals
NBP3-20342-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18608014
View Documents
Novus Biologicals
NBP3-20342-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications

Specifications

S100 calcium binding protein A14
0.2 mg/ml
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer
10mM PBS with 0.05% BSA
57402
IgG2a κ
Protein A or G purified
S100A14/7401
ELISA, Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human
protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114
Recombinant full-length human S100 calcium binding protein A14 protein
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.