missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Verapamilic Acid, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | (S)-(-)-Verapamilic Acid |
| CAS | 36622-24-9 |
| Molecular Formula | C16 H21 N O4 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C16H21NO4/c1-11(2)16(10-17,8-7-15(18)19)12-5-6-13(20-3)14(9-12)21-4/h5-6,9,11H,7-8H2,1-4H3,(H,18,19)/t16-/m0/s1 |
| SMILES | COc1ccc(cc1OC)[C@@](CCC(=O)O)(C#N)C(C)C |
| IUPAC Name | (4S)-4-cyano-4-(3,4-dimethoxyphenyl)-5-methylhexanoic acid |
| Molecular Weight (g/mol) | 291.34 |
| Formula Weight | 291.1471 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction