missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Valsartan-d8 (valine-d8), TRC

Product Code. 30453679
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453679 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30453679 Supplier Toronto Research Chemicals Supplier No. TRCV09575110MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-(-)-Valsartan-d8 (valine-d8)
CAS 1089736-72-0
Molecular Formula C24H21D8N5O3
Quantity 10 mg
Synonym L-Valine-2,3,4,4,4,4',4',4'-d8','N-(1-oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (ACI), N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine-2,3,4,4,4,4',4',4'-d8 (ACI), (2S)-2,3,4,4,4-Pentadeuterio-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]-3-(trideuteriomethyl)butanoic acid, Valsartan D8 (Valine D8), Valsartan-d8, Valsartan-D8 (L-valine-D8), (S)-(-)-Valsartan-d8 (valine-d8), N-(1-Oxopentyl)-N-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-L-valine-2,3,4,4,4,4',4',4'-d8
InChI Formula InChI=1S/C24H29N5O3/c1-4-5-10-21(30)29(22(16(2)3)24(31)32)15-17-11-13-18(14-12-17)19-8-6-7-9-20(19)23-25-27-28-26-23/h6-9,11-14,16,22H,4-5,10,15H2,1-3H3,(H,31,32)(H,25,26,27,28)/t22-/m0/s1/i2D3,3D3,16D,22D
SMILES [2H]C([2H])([2H])C([2H])(C([2H])([2H])[2H])[C@]([2H])(N(Cc1ccc(cc1)c2ccccc2c3nnn[nH]3)C(=O)CCCC)C(=O)O
IUPAC Name (2S)-2,3,4,4,4-pentadeuterio-2-[pentanoyl-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]amino]-3-(trideuteriomethyl)butanoic acid
Molecular Weight (g/mol) 443.58
Formula Weight 443.2773
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.