missing translation for 'onlineSavingsMsg'
Learn More

(S)-Timolol Maleate, MedChemExpress

Product Code. 16477266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477266 100 mg 100mg
16471246 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477266 Supplier Medchem Express Supplier No. HY17380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. (S)-Timolol Maleate is widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. (S)-Timolol Maleate can be used for hypertension, angina pectoris and myocardial infarction.

(S)-Timolol Maleate (L-714,465 Maleate) is a non-cardioselective hydrophilic β-adrenoceptor blocker. (S)-Timolol Maleate is widely used as standard medication for intraocular pressure (glaucoma) by preventing the production of aqueous humor. (S)-Timolol Maleate can be used for hypertension, angina pectoris and myocardial infarction.

Specifications

Chemical Name or Material (S)-Timolol Maleate
CAS 26921-17-5
Color White
Purity Grade Notes Research
Molecular Formula C17H28N4O7S
Quantity 100 mg
Synonym L-714,465 Maleate MK 950
Solubility Information DMSO : 100 mg/mL (231.22 mM; Need ultrasonic) ∣H2O : 50 mg/mL (115.61 mM; Need ultrasonic)
SMILES O[C@H](COC1=NSN=C1N2CCOCC2)CNC(C)(C)C.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 432.49
Formula Weight 432.49
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.