missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydro-2-furoic acid, 99% (98% ee), Thermo Scientific™

Catalog Number p-3830598
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
87392-07-2
C5H8O3
116.12
MFCD00272639
UJJLJRQIPMGXEZ-BYPYZUCNSA-N
s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
446666
CHEBI:45960
(2S)-oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O
This item is not returnable. View return policy
87392-07-2
C5H8O3
116.12
MFCD00272639
UJJLJRQIPMGXEZ-BYPYZUCNSA-N
s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
446666
CHEBI:45960
(2S)-oxolane-2-carboxylic acid
C1CC(OC1)C(=O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.2100g/mL
Boiling Point 244.0°C (736.0 mmHg)
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4570 to 1.4590
Specific Gravity 1.21
Solubility Information Solubility in water: soluble.
Formula Weight 116.12
Percent Purity 99% (98% ee)
Chemical Name or Material (S)-(-)-Tetrahydro-2-furoic acid, 98% e.e.
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-2-Tetrahydrofuroic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'