missing translation for 'onlineSavingsMsg'
Learn More

(S)-tert-Leucinol, 95%

Product Code. 10706953 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10706953

Brand: Thermo Scientific Acros 377532500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 112245-13-3 | C6H16NO | 118.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112245-13-3
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00192250
InChI Key JBULSURVMXPBNA-RXMQYKEDSA-O
Synonym l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
PubChem CID 2734079
SMILES CC(C)(C)[C@H]([NH3+])CO

Specifications

Infrared Spectrum Authentic
Color White or Colorless
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Specific Rotation +34° to +40° (25°C, 589nm) (c=1.5, EtOH)
Quantity 250 mg
Physical Form Crystalline Low Melting Mass or Liquid
Assay Percent Range 95%
Linear Formula (CH3)3CCH(NH2)CH2OH
IUPAC Name (2S)-2-amino-3,3-dimethylbutan-1-ol
Formula Weight 117.19
Percent Purity ≥94%
Chemical Name or Material (S)-tert-Leucinol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-tert-Leucinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.