missing translation for 'onlineSavingsMsg'
Learn More

(S)-Tedizolid, MedChemExpress

Product Code. 16492337
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492337 50 mg 50mg
16476208 10 mg 10mg
16418807 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492337 Supplier Medchem Express Supplier No. HY14855A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Tedizolid is the S-enantiomer of Tedizolid. Tedizolid is a novel oxazolidinone with activity against Gram-positive pathogens. (S)-Tedizolid is the less active isomer.

(S)-Tedizolid is the S-enantiomer of Tedizolid. Tedizolid is a novel oxazolidinone with activity against Gram-positive pathogens. (S)-Tedizolid is the less active isomer.

Specifications

Chemical Name or Material (S)-Tedizolid
CAS 1431699-67-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H15FN6O3
Quantity 50 mg
Synonym (S)-TR 700 (S)-DA 7157
Solubility Information DMSO : ≥ 100 mg/mL (270.02 mM)
SMILES O=C1O[C@H](CO)CN1C2=CC=C(C3=CC=C(C4=NN(C)N=N4)N=C3)C(F)=C2
Molecular Weight (g/mol) 370.34
Formula Weight 370.34
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.