missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (S)-(-)-Sulpiride

Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
(S)-(-)-Sulpiride is an active enantiomer. Selective D2-like dopamine antagonist (Ki values are ∼ 0.015, ∼ 0.013, 1, ∼ 45 and ∼ 77 μM at D2, D3, D4, D1 and D5 receptors respectively).
Racemate also available.
Specifications
Specifications
| CAS | 23672-07-3 |
| Quantity | 100 mg |
| Molecular Formula | C15H23N3O4S |
| Formula Weight | 341.42 |
| Purity | >99% |
| Molecular Weight (g/mol) | 341.426 |
| Chemical Name or Material | (S)-(-)-Sulpiride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction