missing translation for 'onlineSavingsMsg'
Learn More

(S)-Styrene oxide, 94%

Product Code. 15294764 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15294764 25 g Glass bottle 25g
15150967 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15294764 Supplier Thermo Scientific Acros Supplier No. 449450250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20780-54-5 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20780-54-5
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00064310,MFCD00066210
InChI Key AWMVMTVKBNGEAK-MRVPVSSYSA-N
Synonym s-styrene oxide, s-2-phenyloxirane, 2s-2-phenyloxirane, s-phenyloxirane, s-epoxyethyl benzene, s-+-styrene oxide, styrene oxide, s, unii-av5p894c84, ccris 4094
PubChem CID 114946
ChEBI CHEBI:51014
IUPAC Name (2S)-2-phenyloxirane
SMILES C1O[C@H]1C1=CC=CC=C1

Specifications

Melting Point -37.0°C
Density 1.0510g/mL
Boiling Point 192.0°C to 194.0°C
Flash Point 79°C
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.051
Solubility Information Solubility in water: practically insoluble
Formula Weight 120.15
Percent Purity 94%
Chemical Name or Material (S)-Styrene oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-Styrene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.