missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-Hydrobenzoin, 98+%

Product Code. 15428999
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428999 25 g 25g
15418999 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428999 Supplier Thermo Scientific Alfa Aesar Supplier No. L14082.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2325-10-2 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2325-10-2
MDL Number MFCD00064255

Specifications

Melting Point 148°C to 150°C
Color White
Molecular Formula C14H14O2
Quantity 25 g
Beilstein 2330888
Merck Index 14,4777
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol) 214.26
PubChem CID 853020
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material (S,S)-(-)-Hydrobenzoin
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.