missing translation for 'onlineSavingsMsg'
Learn More

(S,S)Dinitroso Ethambutol, TRC

Product Code. 30357604
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357604 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357604 Supplier Toronto Research Chemicals Supplier No. TRCD26285025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Dinitrosoethambutol
CAS 2792161-95-4
Molecular Formula C10 H22 N4 O4
Quantity 25 mg
Synonym N,N'-Dinitrosoethambutol
InChI Formula InChI=1S/C10H22N4O4/c1-3-9(7-15)13(11-17)5-6-14(12-18)10(4-2)8-16/h9-10,15-16H,3-8H2,1-2H3/t9-,10-/m0/s1
SMILES CC[C@@H](CO)N(CCN(N=O)[C@@H](CC)CO)N=O
IUPAC Name N-[(2S)-1-hydroxybutan-2-yl]-N-[2-[[(2S)-1-hydroxybutan-2-yl]-nitrosoamino]ethyl]nitrous amide
Molecular Weight (g/mol) 262.3061
Formula Weight 262.1641
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.