missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine, 97%, Thermo Scientific Chemicals

Product Code. 11995921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995921 100 mg Glass bottle 100mg
11905931 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11995921 Supplier Thermo Scientific Acros Supplier No. 437281000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163169-29-7
Molecular Formula C28H28FeNOP
Molecular Weight (g/mol) 481.36
MDL Number MFCD15144869
InChI Key FELCHLOAEMNOQM-KISGOYOSNA-N
Synonym (S)[(Sp)-2-(Diphenylphosphino)ferrocenyl]-4-isopropyloxazoline
IUPAC Name (S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
SMILES [Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 162.0°C to 165.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 481.35
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S,S)-[2-(4′-Isopropyloxazolin-2′yl)ferrocenyl]diphenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.