missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

Product Code. 11381889
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381889 1 g 1g
15487535 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381889 Supplier Alfa Aesar™ Supplier No. H25993.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1052707-27-3
Molecular Formula C22H22Cl2N2
Molecular Weight (g/mol) 385.332
MDL Number MFCD09265324
InChI Key SHNGCXWOHADIKG-IXOXMDGESA-N
Synonym 1s, 2s-1,2-di-1-naphthyl-ethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthylethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthalenyl-1,2-ethanediaminehydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine dihydrochloride, s, s-1,2-bis 1-naphthyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis naphthalen-1-yl ethane-1,2-diamine dihydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-di 1-naphthyl-1,2-ethanediamine dihydrochloride, ee
PubChem CID 57468034
IUPAC Name (1S,2S)-1,2-dinaphthalen-1-ylethane-1,2-diamine;dihydrochloride
SMILES C1=CC=C2C(=C1)C=CC=C2C(C(C3=CC=CC4=CC=CC=C43)N)N.Cl.Cl

Specifications

Melting Point >200°C
Quantity 1 g
Optical Rotation 235° (c=1 in Water)
Formula Weight 385.35
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.