missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-1,2-Bis(4-nitrophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Product Code. 11425760
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425760 1 g 1g
15425095 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425760 Supplier Thermo Scientific Alfa Aesar Supplier No. H25970.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1052707-07-9
Molecular Formula C14H16Cl2N4O4
Molecular Weight (g/mol) 375.206
MDL Number MFCD09265322
InChI Key PDPYGNJVCKPVGM-AXEKQOJOSA-N
Synonym 1s, 2s-1,2-bis 4-nitrophenyl ethylenediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine dihydrochloride, 1r,2r-1,2-bis 4-nitrophenyl ethane-1,2-diaminedihydrochloride, 1s, 2s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, ee
PubChem CID 45925681
IUPAC Name (1S,2S)-1,2-bis(4-nitrophenyl)ethane-1,2-diamine;dihydrochloride
SMILES C1=CC(=CC=C1C(C(C2=CC=C(C=C2)[N+](=O)[O-])N)N)[N+](=O)[O-].Cl.Cl

Specifications

Melting Point 196°C
Quantity 1 g
Optical Rotation −83° (c=1 in Water)
Formula Weight 375.23
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (S,S)-1,2-Bis(4-nitrophenyl)-1,2-ethanediamine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.