missing translation for 'onlineSavingsMsg'
Learn More

(S)-Rasagiline, MedChemExpress

Product Code. 16415998
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16415998 10 mg 10mg
16408617 5 mg 5mg
16403416 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16415998 Supplier Medchem Express Supplier No. HY14200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S)-Rasagiline (TVP1022) is the relatively inactive S-enantiomer form of Rasagiline. Rasagiline is a highly potent selective irreversible MAO inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

(S)-Rasagiline (TVP1022) is the relatively inactive S-enantiomer form of Rasagiline. Rasagiline is a highly potent selective irreversible MAO inhibitor with IC50s of 4.43 nM and 412 nM for rat brain MAO B and A activity, respectively.

Specifications

Chemical Name or Material (S)-Rasagiline
CAS 185517-74-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H13N
Quantity 10 mg
Synonym TVP1022 S-PAI
Solubility Information DMSO : 100 mg/mL (583.98 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (583.98 mM; Need ultrasonic)
SMILES C#CCN[C@H]1CCC2=CC=CC=C21
Molecular Weight (g/mol) 171.24
Formula Weight 171.24
Percent Purity 98.8%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.