missing translation for 'onlineSavingsMsg'
Learn More

(S,R,S)-AHPC-Me, MedChemExpress

Product Code. 16405416
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16405416

Brand: Medchem Express HY112078

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-Me (VHL ligand 2) is the (S,R,S)-AHPC-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC-Me can be used to synthesize ARV-771, a von Hippel-Landau (VHL) E3 ligase-based BET PROTAC degrader. ARV-771 potently degrades BET protein in castration-resistant prostate cancer (CRPC) cells with a DC50 <1 nM.

(S,R,S)-AHPC-Me (VHL ligand 2) is the (S,R,S)-AHPC-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC-Me can be used to synthesize ARV-771, a von Hippel-Landau (VHL) E3 ligase-based BET PROTAC degrader. ARV-771 potently degrades BET protein in castration-resistant prostate cancer (CRPC) cells with a DC50 <1 nM.

Specifications

Chemical Name or Material (S,R,S)-AHPC-Me
CAS 1948273-02-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H32N4O3S
Quantity 100 mg
Synonym VHL ligand 2 E3 ligase Ligand 1A
Solubility Information DMSO : 250 mg/mL (562.32 mM; Need ultrasonic)
SMILES N[C@@H](C(C)(C)C)C(N1[C@H](C(N[C@@H](C)C2=CC=C(C3=C(C)N=CS3)C=C2)=O)C[C@@H](O)C1)=O
Molecular Weight (g/mol) 444.59
Formula Weight 444.59
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.