missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,R,S)-AHPC-Me, MedChemExpress
Description
(S,R,S)-AHPC-Me (VHL ligand 2) is the (S,R,S)-AHPC-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC-Me can be used to synthesize ARV-771, a von Hippel-Landau (VHL) E3 ligase-based BET PROTAC degrader. ARV-771 potently degrades BET protein in castration-resistant prostate cancer (CRPC) cells with a DC50 <1 nM.
Specifications
Specifications
| Chemical Name or Material | (S,R,S)-AHPC-Me |
| CAS | 1948273-02-6 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C23H32N4O3S |
| Quantity | 500 mg |
| Synonym | VHL ligand 2 E3 ligase Ligand 1A |
| Solubility Information | DMSO : 250 mg/mL (562.32 mM; Need ultrasonic) |
| SMILES | N[C@@H](C(C)(C)C)C(N1[C@H](C(N[C@@H](C)C2=CC=C(C3=C(C)N=CS3)C=C2)=O)C[C@@H](O)C1)=O |
| Molecular Weight (g/mol) | 444.59 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?