missing translation for 'onlineSavingsMsg'
Learn More

(S,R,S)-AHPC hydrochloride, MedChemExpress

Product Code. 16480899
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480899 1 g 1g
16474666 100 mg 100mg
16441536 500 mg 500mg
16406287 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480899 Supplier Medchem Express Supplier No. HY101763A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells.

(S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells.

Specifications

Chemical Name or Material (S,R,S)-AHPC hydrochloride
CAS 1448189-80-7
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClN4O3S
Quantity 1 g
Synonym VH032-NH2 hydrochlorideVHL ligand 1 hydrochloride
Solubility Information DMSO : 250 mg/mL (535.31 mM; Need ultrasonic) ∣H2O : 100 mg/mL (214.12 mM; Need ultrasonic)
SMILES O=C([C@H](C[C@@H](O)C1)N1C([C@@H](N)C(C)(C)C)=O)NCC2=CC=C(C(SC=N3)=C3C)C=C2.Cl
Molecular Weight (g/mol) 467.02
Formula Weight 467.02
Percent Purity 95.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.