missing translation for 'onlineSavingsMsg'
Learn More

(S,R,S)-AHPC hydrochloride, MedChemExpress

Product Code. 16441536
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
This item is not returnable. View return policy

Product Code. 16441536

Brand: Medchem Express HY101763A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells.

(S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells.

Specifications

Chemical Name or Material (S,R,S)-AHPC hydrochloride
CAS 1448189-80-7
Color White
Purity Grade Notes Research
Molecular Formula C22H31ClN4O3S
Quantity 500 mg
Synonym VH032-NH2 hydrochlorideVHL ligand 1 hydrochloride
Solubility Information DMSO : 250 mg/mL (535.31 mM; Need ultrasonic) ∣H2O : 100 mg/mL (214.12 mM; Need ultrasonic)
SMILES O=C([C@H](C[C@@H](O)C1)N1C([C@@H](N)C(C)(C)C)=O)NCC2=CC=C(C(SC=N3)=C3C)C=C2.Cl
Molecular Weight (g/mol) 467.02
Formula Weight 467.02
Percent Purity 95.05%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.