missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,R,S)-AHPC-C4-NH2 hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
Description
(S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.
Specifications
Specifications
| Chemical Name or Material | (S,R,S)-AHPC-C4-NH2 hydrochloride |
| CAS | 2245697-83-8 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C27H40ClN5O4S |
| Quantity | 1 g |
| Synonym | VH032-C4-NH2 hydrochloride VHL Ligand-Linker Conjugates 13 E3 ligase Ligand-Linker Conjugates 28 |
| Solubility Information | DMSO : ≥ 109.6 mg/mL (193.58 mM) ∣H2O : ≥ 50 mg/mL (88.31 mM) |
| SMILES | O=C(N[C@@H](C(C)(C)C)C(N1C[C@H](O)C[C@H]1C(NCC2=CC=C(C=C2)C3=C(C)N=CS3)=O)=O)CCCCN.[H]Cl |
| Molecular Weight (g/mol) | 566.16 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction