missing translation for 'onlineSavingsMsg'
Learn More

(S,R,S)-AHPC-C4-NH2 hydrochloride, MedChemExpress

Product Code. 16470146
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
2 g
Unit Size:
100mg
1g
2g
500mg
This item is not returnable. View return policy

Product Code. 16470146

Brand: Medchem Express HY114176

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.

(S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC.

Specifications

Chemical Name or Material (S,R,S)-AHPC-C4-NH2 hydrochloride
CAS 2245697-83-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H40ClN5O4S
Quantity 2 g
Synonym VH032-C4-NH2 hydrochloride VHL Ligand-Linker Conjugates 13 E3 ligase Ligand-Linker Conjugates 28
Solubility Information DMSO : ≥ 109.6 mg/mL (193.58 mM) ∣H2O : ≥ 50 mg/mL (88.31 mM)
SMILES O=C(N[C@@H](C(C)(C)C)C(N1C[C@H](O)C[C@H]1C(NCC2=CC=C(C=C2)C3=C(C)N=CS3)=O)=O)CCCCN.[H]Cl
Molecular Weight (g/mol) 566.16
Formula Weight 566.16
Percent Purity 95.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.