missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals (S,R)-Noscapine

Product Code. 16514224 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16514224 5 g 5g
16524224 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16514224 Supplier Thermo Scientific Chemicals Supplier No. 462710050
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 128-62-1 | C22H23NO7 | 413.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-62-1
Molecular Formula C22H23NO7
Molecular Weight (g/mol) 413.43
InChI Key AKNNEGZIBPJZJG-MSOLQXFVSA-N
IUPAC Name (3S)-6,7-dimethoxy-3-[(5R)-4-methoxy-6-methyl-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-1,3-dihydro-2-benzofuran-1-one
SMILES COC1=CC=C2[C@H](OC(=O)C2=C1OC)[C@@H]1N(C)CCC2=CC3=C(OCO3)C(OC)=C12

Specifications

Melting Point 174.0°C to 177.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: practically insoluble. Other solubilities: solubel in ethanol, chloroform, acetone, glacial, acetic acid
Formula Weight 413.42
Chemical Name or Material (S,R)-Noscapine
compliance-icons
Product Identifier
  • (S,R)-Noscapine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Noscapine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.