missing translation for 'onlineSavingsMsg'
Learn More

(S,R)-cis-Clopidogrel-MP Derivative, TRC

Product Code. 30357868
Change view
Click to view available options
Quantity:
0.25 mg
0.5 mg
Unit Size:
0.25mg
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357868 0.25 mg 0.25mg
30360205 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357868 Supplier Toronto Research Chemicals Supplier No. TRCC5885250.25MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1430373-14-0
Molecular Formula C25H26ClNO6S
Molecular Weight (g/mol) 504
Synonym 1-Methyl-(alphaS,3Z,4R)-3-(carboxymethylene)-alpha-(2-chlorophenyl)-4-[[2-(3-methoxyphenyl)-2-oxoethyl]thio]-1-piperidineacetate
IUPAC Name (2Z)-2-[(4R)-1-[(1S)-1-(2-chlorophenyl)-2-methoxy-2-oxoethyl]-4-[2-(3-methoxyphenyl)-2-oxoethyl]sulfanylpiperidin-3-ylidene]acetic acid
SMILES COC(=O)[C@@H](N1CC[C@@H](SCC(=O)c2cccc(OC)c2)\C(=C/C(=O)O)\C1)c3ccccc3Cl

Specifications

Quantity 0.25 mg
InChI Formula InChI=1S/C25H26ClNO6S/c1-32-18-7-5-6-16(12-18)21(28)15-34-22-10-11-27(14-17(22)13-23(29)30)24(25(31)33-2)19-8-3-4-9-20(19)26/h3-9,12-13,22,24H,10-11,14-15H2,1-2H3,(H,29,30)/b17-13-/t22-,24+/m1/s1
Formula Weight 503.1169
Chemical Name or Material (S,R)-cis-Clopidogrel-MP Derivative
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.