missing translation for 'onlineSavingsMsg'
Learn More

(S)-Quizalofop Ethyl, TRC

Product Code. 30423351
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423351 500 mg -
30408986 50 mg 50mg
30415710 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423351 Supplier Toronto Research Chemicals Supplier No. TRCQ164650500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100646-52-4
Molecular Formula C19H17ClN2O4
Molecular Weight (g/mol) 372.8
Synonym 2-[4-[(6-Chloro-2-quinoxalinyl)oxy]phenoxy]-Ethyl Ester-(S)-Propanoic Acid, (-)-Quizalofop Ethyl
IUPAC Name ethyl (2S)-2-[4-(6-chloroquinoxalin-2-yl)oxyphenoxy]propanoate
SMILES CCOC(=O)[C@H](C)Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3/t12-/m0/s1
Formula Weight 372.0877
Chemical Name or Material (S)-Quizalofop Ethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.