missing translation for 'onlineSavingsMsg'
Learn More

(S)-Purvalanol B, MedChemExpress

Product Code. 16451729
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451729 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451729 Supplier Medchem Express Supplier No. HY18299B
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-Purvalanol B is the S enantiomer of Purvalanol B. Purvalanol B is a cyclin-dependent kinase inhibitor.

(S)-Purvalanol B is the S enantiomer of Purvalanol B. Purvalanol B is a cyclin-dependent kinase inhibitor.

Specifications

Chemical Name or Material (S)-Purvalanol B
Color White
Purity Grade Notes Research
Molecular Formula C20H25ClN6O3
Quantity 1 mg
Synonym (S)-NG 95
Solubility Information DMSO : 76.7 mg/mL (177.18 mM; Need ultrasonic and warming)
SMILES O=C(O)C1=CC=C(NC2=C3N=CN(C(C)C)C3=NC(N[C@H](CO)C(C)C)=N2)C=C1Cl
Molecular Weight (g/mol) 432.9
Formula Weight 432.9
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.