missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Propylene oxide, 99%, Thermo Scientific Chemicals

Product Code. 11426056
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426056 5 g 5g
11436056 25 g 25g
11446056 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426056 Supplier Thermo Scientific Alfa Aesar Supplier No. B22341.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

CAS: 16088-62-3 | C3H6O | 58.08 g/mol

(S)-(-)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used in making the polyurethane plastics. It finds an application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It is also used in model aircraft and surface vehicles as a glow fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-Propylene oxide is widely utilized in the manufacture of polyether polyols and propylene glycol, which is used in making the polyurethane plastics. It finds an application as a fumigant for the sterilization of packaged foods and plastic medical instruments. Combine with ethanol, it used to make biological samples for electron microscopy. It is also used in model aircraft and surface vehicles as a glow fuel.

Solubility
Miscible with water and many organic solvents.

Notes
Highly flammable. Keep away from sources of ignition. The substance may polymerize under the influence of bases, acids and metal chlorides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16088-62-3
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00064312
InChI Key GOOHAUXETOMSMM-VKHMYHEASA-N
Synonym s---propylene oxide, s-propylene oxide, 2s-2-methyloxirane, s-1,2-epoxypropane, --propylene oxide, s-2-methyloxirane, --methyloxirane, s-methyloxirane, s---1,2-epoxypropane
PubChem CID 146262
ChEBI CHEBI:28982
IUPAC Name (2S)-2-methyloxirane
SMILES C[C@H]1CO1

Specifications

Density 0.829
Boiling Point 34°C to 35°C
Flash Point −37°C (−35°F)
Odor Ether-like
Refractive Index 1.367
Quantity 5 g
UN Number UN1280
Beilstein 79765
Merck Index 14,7856
Solubility Information Miscible with water and many organic solvents.
Optical Rotation −14° (Neat)
Formula Weight 58.08
Percent Purity 99%
Chemical Name or Material (S)-(-)-Propylene oxide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-Propylene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 1
  • Germ cell mutagenicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: (S)-1,2-Epoxypropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.