missing translation for 'onlineSavingsMsg'
Learn More

(S)-Pramipexole Dihydrochloride, TRC

Product Code. 30413755
Change view
Click to view available options
Quantity:
5 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30413755 100 mg
30408809 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413755 Supplier Toronto Research Chemicals Supplier No. TRCP700755100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104632-25-9
Molecular Formula C10 H17 N3 S . 2 Cl H
Molecular Weight (g/mol) 284.25
Synonym 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, hydrochloride (1:2), (6S)- (ACI), 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, dihydrochloride, (6S)- (9CI), 2,6-Benzothiazolediamine, 4,5,6,7-tetrahydro-N6-propyl-, dihydrochloride, (S)- (ZCI), Pramipexole dihydrochloride, Pramipexole hydrochloride, SND 19, (6S)-4,5,6,7-Tetrahydro-N6-propyl-2,6-benzothiazolediamine dihydrochloride
IUPAC Name (6S)-6-N-propyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine;dihydrochloride
SMILES Cl.Cl.CCCN[C@H]1CCc2nc(N)sc2C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H17N3S.2ClH/c1-2-5-12-7-3-4-8-9(6-7)14-10(11)13-8;;/h7,12H,2-6H2,1H3,(H2,11,13);2*1H/t7-;;/m0../s1
Formula Weight 283.0677
Chemical Name or Material Pramipexole Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.