missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Piperazine-2-carboxylic acid dihydrochloride, 98+%

Product Code. 11452200
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452200 5 g 5g
15477745 250 mg 250mg
11442200 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452200 Supplier Thermo Scientific Alfa Aesar Supplier No. H28972.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 158663-69-5 | C5H12Cl2N2O2 | 203.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158663-69-5
Molecular Formula C5H12Cl2N2O2
Molecular Weight (g/mol) 203.06
MDL Number MFCD01632104,MFCD03840829
InChI Key WNSDZBQLMGKPQS-UHFFFAOYNA-N
Synonym s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
PubChem CID 2734260
IUPAC Name (2S)-piperazine-2-carboxylic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)C1CNCCN1

Specifications

Melting Point ∽280°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Optical Rotation −5° (c=1 in Water)
Formula Weight 203.07
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-Piperazine-2-carboxylic acid dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.