missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ S-PHOENIX-90 Light Gray Safety Storage Cabinets with Auto Closing Doors

Product Code. 15625747 Shop All asecos Products
Click to view available options
Load Bearing Capacity:
Drawers 60 kg, Max load 600 kg
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15625747

Brand: Asecos™ 3011900133726

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Safety storage cabinets with folding doors for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.

  • Optimal developed door technology for easy operation
  • Air-dampened door buffer ensures soft and low-wear closings
  • Available in a range of widths and heights
  • Space-saving folding doors – ideal for use in case of narrow passage widths in front of the cabinet
  • Convenient handling – door operation with minimal effort
  • Continuous anti-trapping door strips on the folding doors offer maximum protection during operation
  • Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
  • Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
  • Fire resistance of 90 minutes
  • Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system
  • Some models with serial automatic door closing which releases arrested doors after approx. 60 seconds with an acoustic and visual signal before closing
  • One-hand operation for double-door cabinets

Specifications

Certifications/Compliance EN 14470-1, TRGS 510
Configuration 6 x 60 kg/29 L powder-coated sheet steel Drawers
Dimensions (D x W x H) 616 x 1196 x 1968 mm
Door Style Auto-close
For Use With (Application) Storage of flammable hazardous materials
Load Bearing Capacity Drawers 60 kg, Max load 600 kg
No. of Doors 2
Surface Coating Powder-coated
Weight (Metric) 527 kg
Color Light Gray
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.