missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Pantolactone, TRC

Product Code. 30420258
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420258 1 g
30410626 250 mg
30420606 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420258 Supplier Toronto Research Chemicals Supplier No. TRCP1826001G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5405-40-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
Synonym L-(+)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (3S)-Dihydro-3-hydroxy-4,4-dimethyl-2(3H)-furanone, (+)-2-Hydroxy-3,3-dimethyl-gamma-butyrolactone, (+)-Pantolactone, (+)-Pantoyl Lactone, (S)-(+)-Pantolactone, L-(+)-Pantolactone, L-(+)-Pantoyl Lactone, L-(+)-alpha-Hydroxy-beta,beta-dimethyl-gamma-butyrolactone, L-Pantolactone, L-Pantoyl Lactone
IUPAC Name (3S)-3-hydroxy-4,4-dimethyloxolan-2-one
SMILES CC1(C)COC(=O)[C@H]1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H10O3/c1-6(2)3-9-5(8)4(6)7/h4,7H,3H2,1-2H3/t4-/m1/s1
Formula Weight 130.063
Chemical Name or Material (S)-(+)-Pantolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.