missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Nicotine, TRC

Product Code. 17987749
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17987749 5 mL 5mL
17927159 25 mL 25mL
17909269 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987749 Supplier Toronto Research Chemicals Supplier No. TRCN4124505ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10 H14 N2
Molecular Weight (g/mol) 162.23
Synonym Nicotine (8CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, (S)-, 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine, (-)-(S)-Nicotine, (-)-3-(1-Methyl-2-pyrrolidyl)pyridine, (-)-Nicotine, (-)-β-Pyridyl-α-N-methylpyrrolidine, (S)-(-)-Nicotine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-Nicotine, Exodus, Flux Maag, Habitrol, L-Nicotine, NSC 5065, Nicabate, Nicoderm, Nicolan, Niconil, Nicopatch, Nicorette, Nicotell TTS, Nicotin, Nicotinell, Nicotrol, Nikofrenon, Niquitin, Tabazur, XL All Insecticide, l-Nicotine
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCC[C@H]1c2cccnc2

Specifications

Quantity 5 mL
InChI Formula InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1
Formula Weight 162.1157
Percent Purity >95%
Chemical Name or Material Nicotine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.