missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Nicotine, 98+%

Product Code. p-7082113
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11409563

Brand: Thermo Scientific Alfa Aesar A12398.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Nicotine, CAS # 54-11-5, has been shown in laboratory studies to alleviate smoking habits. It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine. | CAS: 54-11-5 | C10H14N2 | 162.236 g/mol

(S)-(-)-Nicotine is utilized as a therapeutic agent to eliminate smoking with the damage it does to health. It is widely used as a drug to enhance the memory or other cognition functions, which is second to caffeine. Lesser levels of nicotine given in the form of gum, may help to eliminate smoking.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
• It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine
Applications
• L-Nicotine has been shown in studies to alleviate smoking habits
• It has also been shown to enhance cognitive functions such as memory
• Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-11-5
Molecular Formula C10H14N2
MDL Number MFCD00006369
InChI Key SNICXCGAKADSCV-JTQLQIEISA-N
Synonym nicotine, l-nicotine, --nicotine, s-nicotine, habitrol, s-3-1-methylpyrrolidin-2-yl pyridine, nicoderm, nicotrol, s---nicotine, nicoderm cq
PubChem CID 89594
ChEBI CHEBI:17688
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine

Specifications

Melting Point °C to 79°C
Density 1.017
Boiling Point 245°C to 248°C
Flash Point 101°C (213°F)
Assay Percent Range 98+%
Refractive Index 1.5265
Quantity 25 g
UN Number UN1654
Beilstein 82109
Sensitivity Air and light sensitive; Hygroscopic
Merck Index 14,6524
Solubility Information Miscible with water,alcohol,ether and light petroleum.
SMILES CN1CCCC1C2=CN=CC=C2
Molecular Weight (g/mol) 162.236
Formula Weight 162.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-Nicotine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-Nicotine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : Nicotine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.