missing translation for 'onlineSavingsMsg'
Learn More

(S)-Naproxen, TRC

Product Code. 30133487
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133487 5 g 5g
18083510 10 g 10g
18072910 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133487 Supplier Toronto Research Chemicals Supplier No. TRCN3775205G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22204-53-1
Molecular Formula C14 H14 O3
Molecular Weight (g/mol) 230.26
Synonym 2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (αS)- (9CI), 2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (+)- (8CI), 2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, (S)-, (+)-(S)-Naproxen, (+)-2-(6-Methoxy-2-naphthyl)propionic acid, (+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (+)-Naproxen, (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-(+)-Naproxen, (S)-(+)-Naproxene, (S)-2-(6-Methoxy-2-naphthyl)propanoic acid, (S)-2-(6-Methoxy-2-naphthyl)propionic acid, (S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, (S)-Naproxen, Apo-Naproxen, Bonyl, CG 3117, Diocodal, Dysmenalgit, Equiproxen, Floginax, Laraflex, Laser, MNPA, Naixan, Napren, Naprium, Naprius, Naprosyn, Naprosyne, Naproxen, Naprux, Naxen, Nycopren, Panoxen, Prexan, Proxen, Proxine, RS 3540, Reuxen, Veradol, Xenar, d-2-(6-Methoxy-2-naphthyl)propionic acid, d-Naproxen
IUPAC Name (2S)-2-(6-methoxynaphthalen-2-yl)propanoic acid
SMILES COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C14H14O3/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10/h3-9H,1-2H3,(H,15,16)/t9-/m0/s1
Formula Weight 230.0943
Percent Purity >95%
Chemical Name or Material Naproxen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.