missing translation for 'onlineSavingsMsg'
Learn More

(S)-Naproxen Sodium Salt, TRC

Product Code. 30133820
Change view
Click to view available options
Quantity:
50 mg
5 g
50 g
Unit Size:
50g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133820 50 g 50g
30134346 50 mg 50mg
30005637 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133820 Supplier Toronto Research Chemicals Supplier No. TRCN37752550G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26159-34-2
Molecular Formula C14 H13 O3 . Na
Molecular Weight (g/mol) 252.24
Synonym Naproxen Sodium, (αS)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt (1:1), (S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt, (αS)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt, L-(-)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt, (2S)-2-(6-Methoxy-2-naphthyl)propanoic acid sodium salt, Aflaxen, Agilxen, Aleve, Anaprox, Anaprox DS, Anax, Apranax, Apraxin, Apronax, Axer, Dafloxen, Duk-F, Flanax, Flanax Forte, Gynestrel, Lefaine, Licorax, Miranax, Naprelan, Naprogesic, Naprosyn sodium, Naproxen sodium, Narocin, Primeral, RS 3560, Sodium (+)-6-methoxy-2-naphthalenepropionate, Sodium d-2-(6-methoxy-2-naphthyl)propionate, Sodium naprosyn, Sodium naproxen, Sutolin, Synflex, Synflex Forte, Uniflam, Xenobid
IUPAC Name sodium;(2S)-2-(6-methoxynaphthalen-2-yl)propanoate
SMILES [Na+].COc1ccc2cc(ccc2c1)[C@H](C)C(=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1
Formula Weight 252.0762
Chemical Name or Material Naproxen Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.