missing translation for 'onlineSavingsMsg'
Learn More

S-n-Propyl thioacetate, 98+%, Thermo Scientific™

Product Code. 11464937
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464937 5 g 5g
11474937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464937 Supplier Thermo Scientific Alfa Aesar Supplier No. B21875.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2307-10-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD00039937
InChI Key SBWFWBJCYMBZEY-UHFFFAOYSA-N
Synonym s-propyl thioacetate, propyl thioacetate, ethanethioic acid, s-propyl ester, propyl thiolacetate, s-n-propyl thioacetate, acetic acid, thio-, s-propyl ester, unii-y7284nz6ep, 1-propylsulfanyl ethanone, n-propyl thio acetate, fema no. 3385
PubChem CID 61295
IUPAC Name S-propyl ethanethioate
SMILES CCCSC(C)=O

Specifications

Density 0.971
Boiling Point 137°C to 139°C
Refractive Index 1.46
Quantity 5 g
UN Number UN3272
Beilstein 1740765
Formula Weight 118.2
Percent Purity ≥98%
Chemical Name or Material S-n-Propyl thioacetate
compliance-icons
Product Identifier
  • S-n-Propyl thioacetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: S-n-Propyl thioacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.