missing translation for 'onlineSavingsMsg'
Learn More

(+)-S-N-Methylcoclaurine Hydrochloride, TRC

Product Code. 30138541
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138541 50 mg 50mg
30007404 10 mg 10mg
30012534 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138541 Supplier Toronto Research Chemicals Supplier No. TRCM29432550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1087708-38-0
Molecular Formula C18H21NO3 . HCl
Molecular Weight (g/mol) 335.8
Synonym 1,2,3,4-Tetrahydro-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-7-isoquinolinol Hydrochloride (1:1), (S)-2-Methylcoclaurine Hydrochloride, N-Methyl-coclaurine Hydrochloride, (+)-(S)-N-Methylcoclaurine Hydrochloride, (+)-N-Methylcoclaurine Hydrochloride, N-Methylcoclaurine Hydrochloride, S-N-Methylcoclaurine Hydrochloride
IUPAC Name (1S)-1-[(4-hydroxyphenyl)methyl]-6-methoxy-2-methyl-3,4-dihydro-1H-isoquinolin-7-ol;hydrochloride
SMILES Cl.COc1cc2CCN(C)[C@@H](Cc3ccc(O)cc3)c2cc1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H21NO3.ClH/c1-19-8-7-13-10-18(22-2)17(21)11-15(13)16(19)9-12-3-5-14(20)6-4-12;/h3-6,10-11,16,20-21H,7-9H2,1-2H3;1H/t16-;/m0./s1
Formula Weight 335.1288
Chemical Name or Material (+)-S-N-Methylcoclaurine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.