missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-3-Thienylalanine, 95%, 98% ee, ACROS Organics™

Product Code. 10088751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10088751 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10088751 Supplier Thermo Scientific Acros Supplier No. 361445000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 186320-06-9
CAS Min % 94.0
CAS Max % 100.0
Color White
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C22H19NO4S
MDL Number MFCD00151916
Quantity 500 mg
Synonym fmoc-l-3-thienylalanine, fmoc-3-ala 3-thienyl-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-beta-3-thienyl-ala-oh, fmoc-3-3-thienyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-d-3-3-thienyl-alanine, fmoc-l-3-3-thienyl alanine, n-fmoc-3-4-thienyl-l-alanine, fmoc-3 thg-oh
InChI Key LSBZJMRHROCYGY-FQEVSTJZSA-N
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid
Molecular Weight (g/mol) 393.457
PubChem CID 2734455
Formula Weight 393.46
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-3-Thienylalanine, 95%, (98% e.e.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.