missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-FMOC-2-Naphthylalanine, 95%, 98% e.e., ACROS Organics™

Product Code. 10088691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10088691 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10088691 Supplier Thermo Scientific Acros Supplier No. 361400010
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 112883-43-9
CAS Min % 94.0
CAS Max % 100.0
Color White to Yellow
Enantiomeric Excess 97.5% min. (HPLC)
Infrared Spectrum Authentic
Assay Percent Range 95%
Molecular Formula C28H22NO4
MDL Number MFCD00144886
Quantity 1 g
Synonym fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347
InChI Key JYUTZJVERLGMQZ-SANMLTNESA-M
SMILES [O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol) 436.49
PubChem CID 2734452
Formula Weight 437.39
Percent Purity ≥94%
Physical Form Powder
Chemical Name or Material (S)-N-FMOC-2-Naphthylalanine, 95%, (98% e.e.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.