missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Propargylglycine, 95%

Product Code. p-70083 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10018721 500 mg Glass bottle 500mg
10344162 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10018721 Supplier Thermo Scientific Acros Supplier No. 361805000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 63039-48-5 | C10H15NO4 | 213.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63039-48-5
Molecular Formula C10H15NO4
Molecular Weight (g/mol) 213.23
MDL Number MFCD01320855
InChI Key AMKHAJIFPHJYMH-ZETCQYMHSA-N
Synonym boc-l-propargylglycine, s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, boc-pra-oh, boc-propargyl-gly-oh, n-boc-2-propargyl-l-glycine, 2s-2-tert-butoxycarbonylamino pent-4-ynoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, boc-propargylglycine, boc-l-propargyl glycine
PubChem CID 2734488
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-ynoic acid
SMILES CC(C)(C)OC(=O)NC(CC#C)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 85.5°C
Color White to Yellow
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: soluble.
Formula Weight 213.23
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-BOC-Propargylglycine, 95%, (98% e.e.)
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-BOC-Propargylglycine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.