missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Prolinal, 97%

Product Code. 10063433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063433 5 g 5g
10344552 1 g 1g
11916631 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10063433 Supplier Thermo Scientific Acros Supplier No. 368290050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 69610-41-9 | C10H17NO3 | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69610-41-9
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD00274186,MFCD00801206
InChI Key YDBPZCVWPFMBDH-QMMMGPOBSA-N
Synonym n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
PubChem CID 2734674
IUPAC Name tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC[C@H]1C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.0630g/mL
Boiling Point 211.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630
Quantity 5 g
Specific Gravity 1.063
Specific Rotation Condition −97.5° (20°C c=1,CHCl3)
Specific Rotation − 97.50
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material (S)-N-BOC-Prolinal, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-BOC-Prolinal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.