missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Prolinal, 97%, Thermo Scientific Chemicals

Catalog Number 10344552
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
69610-41-9
C10H17NO3
199.25
MFCD00274186,MFCD00801206
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC[C@H]1C=O
This item is not returnable. View return policy
69610-41-9
C10H17NO3
199.25
MFCD00274186,MFCD00801206
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC[C@H]1C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.0630g/mL
Boiling Point 211.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.4630
Quantity 1 g
Specific Gravity 1.063
Specific Rotation Condition −97.5° (20°C c=1,CHCl3)
Specific Rotation − 97.50
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in chloroform
Formula Weight 199.25
Percent Purity 97%
Chemical Name or Material (S)-N-BOC-Prolinal, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-BOC-Prolinal
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'