missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Allylglycine, 95%, Thermo Scientific Chemicals

Product Code. 10320244 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10320244 5 g Glass Bottle 5g
10176663 500 mg Glass Bottle 500mg
10039692 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10320244 Supplier Thermo Scientific Acros Supplier No. 390100050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90600-20-7 | C10H17NO4 | 215.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90600-20-7
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD01320851
InChI Key BUPDPLXLAKNJMI-ZETCQYMHSA-N
Synonym s-2-tert-butoxycarbonyl amino pent-4-enoic acid, s-n-boc-allylglycine, boc-l-allylglycine, boc-alpha-allyl-l-gly, 4-pentenoic acid, 2-1,1-dimethylethoxy carbonyl amino-, 2s, s-2-tert-butoxycarbonylamino pent-4-enoic acid, 2s-2-tert-butoxycarbonyl amino pent-4-enoic acid, boc-l-alpha-allyl-gly, boc--allyl-l-gly, rarechem bk pt 0250
PubChem CID 2734487
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pent-4-enoic acid
SMILES CC(C)(C)OC(=O)NC(CC=C)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Specific Rotation + 10.50
Specific Rotation Condition +10.50° (20°C c=1,methanol)
Quantity 5 g
Formula Weight 215.25
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material (S)-N-BOC-Allylglycine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.