missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-2-Amino-3-phenylpropyl cyanide, 97+%, Thermo Scientific™

Product Code. 10065023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10065023 5 g Glass Bottle 5g
10548273 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10065023 Supplier Thermo Scientific Acros Supplier No. 374520050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 172695-25-9
Molecular Formula C15H20N2O2
Molecular Weight (g/mol) 260.34
MDL Number MFCD06658950
InChI Key BEJMQXQXIQDKCZ-CYBMUJFWSA-N
Synonym s-tert-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-n-boc-2-amino-3-phenylpropyl cyanide, tert-butyl n-2s-1-cyano-3-phenylpropan-2-yl carbamate, s-t-butyl 1-cyano-3-phenylpropan-2-yl carbamate, s-3-tert-butoxycarbonylamino-4-phenylbutanenitrile, tert-butyl s-1-cyano-3-phenylpropan-2-yl carbamate, tert-butyl 2s-1-cyano-3-phenylpropan-2-yl carbamate, 5-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester, s-1-benzyl-2-cyano-ethyl-carbamic acid tert-butyl ester
PubChem CID 2734830
IUPAC Name tert-butyl N-[(2S)-1-cyano-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CC#N)CC1=CC=CC=C1

Specifications

Melting Point 120°C to 125°C
CAS Min % 97.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5CH2CH(CH2CN)NHCO2C(CH3)2
Quantity 5 g
Formula Weight 260.34
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material (S)-N-BOC-2-Amino-3-phenylpropyl cyanide
Show More Show Less
compliance-icons
Product Identifier
  • (S)-N-BOC-2-Amino-3-phenylpropyl cyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.