missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 15426506
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78879-20-6
Molecular Formula C15H18NO4
Molecular Weight (g/mol) 276.31
MDL Number MFCD00143845
InChI Key HFPVZPNLMJDJFB-LBPRGKRZSA-M
Synonym boc-tic-oh, s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-n-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, boc-l-tic-oh, s-2-boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, n-boc-l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-2-tert-butoxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid, 3s-2-2-methylpropan-2-yl oxycarbonyl-3,4-dihydro-1h-isoquinoline-3-carboxylic acid
PubChem CID 664088
IUPAC Name (3S)-2-[(2-methylpropan-2-yl)oxycarbonyl]-3,4-dihydro-1H-isoquinoline-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CC2=CC=CC=C2C[C@H]1C([O-])=O

Specifications

Color White
Quantity 1 g
Beilstein 4297114
Formula Weight 277.32
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'