missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine, 98%, Thermo Scientific™

Product Code. 11497987
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497987 5 g 5g
11487987 1 g 1g
11407997 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497987 Supplier Thermo Scientific Alfa Aesar Supplier No. A16655.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69632-31-1
Molecular Formula C15H13N3O5
Molecular Weight (g/mol) 315.29
MDL Number MFCD00064498
InChI Key ABEVDCGKLRIYRW-JTQLQIEISA-N
Synonym 3,5-dinitro-n-1s-1-phenylethyl benzamide, s-+-3,5-dinitro-n-1-phenylethyl benzamide, s-+-n-3,5-dinitrobenzoyl-1-phenylethylamine, n-s-1-phenylethyl-3,5-dinitrobenzamide, benzamide,3,5-dinitro-n-1s-1-phenylethyl, b-+-n-3,5-dinitrobenzoyl-alpha-methylbenzylamine, s-+-n-3,5-dinitrobenzoyl-, a-methylbenzylamine, 3-hydroxyazonoyl-5-1s-1-phenylethyl carbamoyl phenyl azinic acid
PubChem CID 5712038
IUPAC Name 3,5-dinitro-N-[(1S)-1-phenylethyl]benzamide
SMILES C[C@H](NC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O)C1=CC=CC=C1

Specifications

Melting Point 158°C to 161°C
Quantity 5 g
Formula Weight 315.28
Percent Purity 98%
Chemical Name or Material (S)-(+)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.