missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™

Product Code. 10234911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234911 10 g Glass bottle 10g
10461202 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234911 Supplier Thermo Scientific Acros Supplier No. 338830100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 66849-29-4
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.23
MDL Number MFCD01862172
InChI Key GXIWMXAAPLZOBY-VIFPVBQESA-N
Synonym s-2-1-phenylethyl amino ethanol, 2-1s-1-phenylethyl amino ethan-1-ol, 2-1s-1-phenylethyl amino ethanol, s---n-2-hydroxyethyl-alpha-phenylethylamine, 2-s-alpha-methylbenzyl amino ethanol, s-n-2-hydroxyethyl-a-phenylethylamine, ethanol,2-1s-1-phenylethyl amino, s-+-n-2-hydroxyethyl-phenylethylamine, s---n-2-hydroxyethyl-alpha-methylbenzylamine
PubChem CID 2733847
IUPAC Name 2-[[(1S)-1-phenylethyl]amino]ethanol
SMILES CC(C1=CC=CC=C1)NCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NH(CH2)2OH
Refractive Index 1.5320 to 1.5350
Quantity 10 g
Specific Rotation Condition − 44.00 (22.00°C c=1,C2H5OH)
Specific Rotation − 44.00
Formula Weight 165.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-N-(2-Hydroxyethyl)-a-phenylethylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.