missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Modafinic Acid, TRC

Product Code. 17960395
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17960395 5 mg 5mg
17931135 25 mg 25mg
17952955 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17960395 Supplier Toronto Research Chemicals Supplier No. TRCM4824755MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112111-44-1
Molecular Formula C15 H14 O3 S
Molecular Weight (g/mol) 274.33
Synonym Acetic acid, 2-[(S)-(diphenylmethyl)sulfinyl]-, Acetic acid, [(S)-(diphenylmethyl)sulfinyl]- (9CI), 2-[(S)-(Diphenylmethyl)sulfinyl]acetic acid, (S)-(+)-Modafinil acid, [(S)-(Diphenylmethyl)sulfinyl]acetic acid
IUPAC Name 2-[(S)-benzhydrylsulfinyl]acetic acid
SMILES OC(=O)C[S@](=O)C(c1ccccc1)c2ccccc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H14O3S/c16-14(17)11-19(18)15(12-7-3-1-4-8-12)13-9-5-2-6-10-13/h1-10,15H,11H2,(H,16,17)/t19-/m0/s1
Formula Weight 274.0664
Chemical Name or Material (S)-(+)-Modafinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.