missing translation for 'onlineSavingsMsg'
Learn More

(S)-ML753286, MedChemExpress

Product Code. 16494296
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494296 100 mg 100mg
16492566 25 mg 25mg
16455727 5 mg 5mg
16439556 50 mg 50mg
16432988 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494296 Supplier Medchem Express Supplier No. HY100390
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (S)-ML753286 is a breast cancer resistance protein (BCRP) inhibitor with an IC50 of 0.6 μM on BCRP efflux transporter.

(S)-ML753286 is a breast cancer resistance protein (BCRP) inhibitor with an IC50 of 0.6 μM on BCRP efflux transporter.

Specifications

Chemical Name or Material (S)-ML753286
CAS 1699720-85-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H25N3O3
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (140.67 mM)
SMILES COC1=CC=C2C(NC3=C2C[C@]4([H])N(C([C@H](C)NC4=O)=O)[C@H]3CC(C)C)=C1
Molecular Weight (g/mol) 355.43
Formula Weight 355.43
Percent Purity 98.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.