missing translation for 'onlineSavingsMsg'
Learn More

Tolvaptan, MedChemExpress

Product Code. 16493267
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16493267

Brand: Medchem Express HY17000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolvaptan is a selective, competitive and orally active vasopressin receptor 2 (V2R) antagonist with an IC50 of 1.28 μM for the inhibition of arginine vasopressin (AVP)-induced platelet aggregation. Tolvaptan induces cell apoposis and affects cell cycle. Tolvaptan can be used for the research of hyponatremia.

Tolvaptan is a selective, competitive and orally active vasopressin receptor 2 (V2R) antagonist with an IC50 of 1.28 μM for the inhibition of arginine vasopressin (AVP)-induced platelet aggregation. Tolvaptan induces cell apoposis and affects cell cycle. Tolvaptan can be used for the research of hyponatremia.

Specifications

Chemical Name or Material Tolvaptan
Physical Form Solid
CAS 150683-30-0
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H25ClN2O3
For Use With (Application) COVID-19-immunoregulation
Synonym OPC-41061
Solubility Information DMSO : ≥ 100 mg/mL (222.75 mM)
SMILES O=C(C1=CC=CC=C1C)NC2=CC=C(C(C)=C2)C(N3CCCC(C4=CC(Cl)=CC=C43)O)=O
Molecular Weight (g/mol) 448.94
Formula Weight 448.94
Percent Purity 98.04%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.